Combustion trays, ceramic, AL24
Supplier: Thermo Fisher Scientific
33187.KFEA 462 EUR
33187.KF
Combustion trays, ceramic, AL24
Boats Combustion Boats
Pure Al₂O₃ Aluminium oxide AL24.
Slightly porous, good resistance to thermal shock, extremely good creep strength.