Pipette tips, OneTouch™
Supplier: Sorenson Bioscience
10310 10370
SORE10310EA 99.7 EUR
SORE10310 SORE10370
Pipette tips, OneTouch™
Pipette Tips
These tips are manufactured with the OneTouch™ dual shot technology. They are made from two different resins: The body is made of rigid PP while the top is made of soft TPE material.
  • Rigid PP body for glass-like clarity; soft TPE top for easy sealing and ejection
  • Tips reduce the necessary sealing and reciprocal ejection force felt with traditional single resin PP tips
  • Soft seal offers a more universal fit: Suitable for use with most pipette brands
  • Ideal for single and multi-channel pipettors
  • Certified RNase/DNase-free, PCR-inhibitor-free and pyrogen-free
  • Top collar is colour-coded by volume