Sample bags, Rigid Safetybag
Supplier: Daklapack
Rigid Safetybag
DAKL460949EA 177 EUR
DAKL460949
Sample bags, Rigid Safetybag
Bags Sample Bags
Safety bag made from PE. Ideal for sending biological samples (category B).
  • Delivered pre-assembled with the top open ready for use with containers or tubes
  • Easy to use: Peel and seal strip ensures a trouble free and quick fluid tight seal
  • Suitable for one time use
  • Easy to open by the recipient by means of an easy open tear notch
The rigid bag is compiled from 3 main components, the fluid tight bag containing a rigid blister inside an absorbing pouch capable of absorbing the contents of a 25 ml tube should any leakage occur.
Certifikace: Complies fully with EU packaging laws and is approved by the BVI (Belgium Packaging Institute).