Moisture analysers
Supplier: Mettler-Toledo
611-0982EA 9940 EUR
611-0982 611-0981
Moisture analysers
Analysers Moisture Analysers
These moisture analysers are based on halogen drying with gold plated reflector technology in a range of formats.
  • Ability to read to 0,001% and 0,01 mg for critical samples
  • Integral printer an option
  • Instrument check
  • Automet swiftly calculates the optimum switch off criteria and can store the parameters as a method
  • 40 methods can be saved in memory
A high specification instrument suitable for use in regulated environments, for both R and D and quality control applications. Method development is facilitated using the Automet feature, which can cut development time by 50%. Sample identification is user specific as there are 4 comment lines available and/or information can be input with a barcode reader.
Informace pro objednávání: For OIML approved models and details on models with integrated printers and accessories, please see the website or contact Avantor sales office.