Jugs, PP, SteriWare®
Supplier: Sampling Systems
8432P-500 8432P-500S 8432P-1000 8432P-1000S
SAMP8432P-500EA 197 EUR
SAMP8432P-500 SAMP8432P-500S SAMP8432P-1000 SAMP8432P-1000S
Jugs, PP, SteriWare®
Bottles Carboys
Made from PP (FDA, EC 1935/2004 and EU 10/2011 conforming).
  • Moulded and packed in a cleanroom
  • BSE/TSE free
  • Full batch traceability
  • Graduation marks at every 10 ml
These range of class leading jugs are available pre-sterilised (gamma irradiated). The ergonomic handle is rounded to make it comfortable to hold and use.
Balení: Individually bagged.