Anti-LAG3 Mouse Monoclonal Antibody (ATTO 488) [clone: 17B4]

Supplier: ProSci
PRSI36-112EA 612 EUR
PRSI36-112
Anti-LAG3 Mouse Monoclonal Antibody (ATTO 488) [clone: 17B4]
Antibodies
Lymphocyte activation gene 3 (LAG-3; CD223) plays an important role in negatively regulating T cell proliferation, function and homeostasis. It is required for maximal natural and induced regulatory T cell (Treg) function. LAG-3 is closely related to the T cell co-receptor CD4 and binds to MHC class II molecules but with a significantly higher affinity than CD4.

Optimal conditions must be determined individually for each application.

Type: Primary
Antigen: LAG3
Clonality: Monoclonal
Clone: 17B4
Conjugation: ATTO 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR