Donepezil Related Compounds, Reference Standard
Supplier: USP
1224981 1224992 1224970
1224981.EA 1610 EUR
1224981. 1224992. 1224970.
Donepezil Related Compounds, Reference Standard
Standards Organic Standards
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifikace: US Pharmacopea Reference Standard
Order Now
SPECIFICATIONS
Description Donepezil Hydrochloride Donepezil Related Compound A Donepezil-N-Oxide
Synonym(s) (E)-2-[(1-benzylpiperidin-4-yl)methylene]-5,6-dimethoxyindan-1-one
Environmentally Preferable
CAS No. 884740-09-4 145546-80-1 120013-84-5
Formula C₂₄H₃₀ClNO₃ C₂₄H₂₇NO₃ C₂₄H₂₉NO₄
UN No. 2811