Dispenser for wipes, WYPALL* 7969
Supplier: Kimberly-Clark
WYPALL*
KIMB7969EA 72.8 EUR
KIMB7969
Dispenser for wipes, WYPALL* 7969
Wipe Dispensers
This versatile dispenser for folded wipes improves hygiene, controls usage, and helps reduce costs. Ideal for quickly and easily dispensing individual WYPALL* wipes with one hand.
  • Quick-fill dispenser in a sleek and contemporary design
  • High-gloss, easy-clean finish (which complies with food safety standards)
  • No dirt or dust traps
  • Comes with different plates to allow multiple dispensing solutions