Dibutoxymethane ≥98.5% (by GC), puriss., Sigma-Aldrich®
Supplier: Merck
Synonyms: 5,7-Dioxahendecane, Formaldehyde Dibutyl Acetal
M-Clarity™ quality level = MQ100
689386-1L 689386-250ML
SIAL689386-1LEA 188 EUR
SIAL689386-1L SIAL689386-250ML
Dibutoxymethane ≥98.5% (by GC), puriss., Sigma-Aldrich®
Dibutoxymethane
Synonyms: Butylal, Dibutoxymethane
Formula: C₉H₁₆O₂
MW: 160.26 g/mol
Density: 0.835 g/ml (20 °C)
Flash Pt: 62 °C
MDL Number: MFCD00059270
CAS Number: 2568-90-3
Order Now
Specification Test Results
Assay ≥98,5% (GC)
Impurities ≤0,2% water
Refractive index n20/D 1,406