Multi-channel pipettes, mechanical, variable volume, Research® plus (General Lab Product)
Supplier: EPPENDORF
3125000.010 3125000.028 3125000.036 3125000.044 3125000.052 3125000.060 3125000.079 3125000.087 3125000.095 3125000.109 3125000.214 3125000.222
613-5903EA 904 EUR
613-5903 613-5906 613-5904 613-5907 613-5905 613-5908 613-6974 613-6975 613-6976 613-6977 613-6978 613-6979
Multi-channel pipettes, mechanical, variable volume, Research® plus (General Lab Product)
Pipettes Micropipettes
Versatile, ergonomic pipette with innovative design, meeting the highest needs in precision and accuracy.
  • Extremely lightweight
  • Innovative re-designed principle of piston-stroke pipetting, piston made of organic polymer Fortron® with great chemical, thermal and mechanical resistance
  • Lowest aspirating, dispensing, blow-out and tip ejection forces
  • Spring loaded retractable nose cone to ensure tight seal and prolong nose cone life, as well as exact tip alignment
  • Individual channel removal for quick and easy servicing
  • Fully autoclavable
Designed and constructed for general laboratory and research applications in Life Science. Typical applications include: Food & beverages, forensic, cosmetics, agriculture, animal studies, chemical analytics, research applications at medical and academic facilities.
Certifikace: Without CE marking.