VWR®, Dip conductivity cells
Supplier: VWR
VWR®
VWRU23198-020EA 266 EUR
VWRU23198-020
VWR®, Dip conductivity cells
Electrodes Conductivity Sensors
Dip Conductivity Cells have a durable epoxy body making them rugged for field use, yet designed for precise and accurate measurements in the laboratory. Ideal for measuring solutions in a test tube or beaker.
  • Built in Thermistor for temperature sensing
  • Made in the USA
Platinum (Pt) cells has a coating of black platinum over the gold (Au) for measurements of high conductivity solutions. Ideal for solutions over 2000 micro Siemens.
All cells come with a 3-month (90 day) limited warranty.