Lumefantrine 98%
Supplier: Thermo Scientific
Synonyms: (9Z)-2,7-Dichloro-9-[(4-chlorophenyl)methylene]-alpha-[(dibutylamino)methyl]-9H-fluoren-4-methanol, Benflumetol, (Z)-2-(Dibutylamino)-1-(2,7-dichloro-9-(4-chlorobenzylidene)-9H-fluoren-4-yl)ethanol
461110050 461110250
ACRO461110050EA 66.9 EUR
ACRO461110050 ACRO461110250
Lumefantrine 98%
Lumefantrine
Formula: C₃₀H₃₂Cl₃NO
MW: 528.94 g/mol
Melting Pt: 136 °C
Storage Temperature: Ambient
MDL Number: MFCD05662268
CAS Number: 82186-77-4
Order Now
Specification Test Results
Appearance Yellow Powder
Infrared spectrum Conforms
Melting point 128 to 134 °C
Loss on drying ≤2 %
HPLC ≥97.5 %