Safety spectacles, PERSPECTA 9000
Supplier: MSA
Perspecta 9000
MSAA10045517EA 9 EUR
MSAA10045517
Safety spectacles, PERSPECTA 9000
Safety Glasses
Incredibly light, resilient and durable protective eyewear for all-day wear. Snug-fitting wrap-around lens provides perfect impact protection around the eyes.
  • Special nose-pads secure glasses to the face, eliminating the problem of slippage while providing a pillow-soft fit
  • Tight-fitting unitary front with dual 9.75 base curvature offers an unobstructed field of vision
  • Revolutionary hard and soft material co-injected temple for improved comfort
Weight: 26 g
Certifikace: EN 166