Microtube membranes, breathable, Breathe-EASIER
Supplier: Diversified Biotech
DIVEBMTM-2000EA 108 EUR
DIVEBMTM-2000
Microtube membranes, breathable, Breathe-EASIER
Seals General Purpose Sealing Films
Breathe-EASIER micropermeable membranes are ideal for cell growth, concentrating samples, and gas exchange. Formatted with an easy to use finger tab, membranes are precut to fit a variety of tube sizes. Membranes are gamma irradiated to ensure sterility and are free of cytotoxins.
  • Microtube cap closures will not bind in centrifuge rotors
  • Convenient scoring makes liner removal quick and easy
  • White non woven rayon membrane rapidly transmits water vapour for accelerated growth