Acetic acid 1 mol/l (1 N) aqueous solution BiopharmaceuticalProduction
Supplier: VWR Chemicals
Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid
6517.9025EA 410 EUR
6517.9025 6517.2500
Acetic acid 1 mol/l (1 N) aqueous solution BiopharmaceuticalProduction
Acetic acid
Formula: H₃CCOOH
MW: 60.05 g/mol
Boiling Pt: 118
Melting Pt: 17
Flash Pt: 38.5
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM