Pentafluorobenzonitrile 99%
Supplier: Thermo Fisher Scientific
|
Danger
|
188340050
188340250
188341000
ACRO188340050EA
28
EUR
ACRO188340050
ACRO188340250
ACRO188341000
Pentafluorobenzonitrile 99%
Pentafluorobenzonitrile
Formula:
C₇F₅N Boiling Pt: 161…162 °C (760 mmHg) Melting Pt: 1…3 °C Density: 1.53 g/cm³ Flash Pt: 29 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001775 CAS Number: 773-82-0 UN: 1993 ADR: 3,III |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4410 to 1.4430 (20°C, 589 nm) |
GC | ≥98.5 % |
HPLC | ≥97.5 % (on dry substance) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...