8-Azahypoxanthine 98%
Supplier: Thermo Fisher Scientific
Warning
202590010 202590050
ACRO202590010EA 160 EUR
ACRO202590010 ACRO202590050
8-Azahypoxanthine 98%
8-Azahypoxanthine
Formula: C₄H₃N₅O
MW: 137.10 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00005804
CAS Number: 2683-90-1
Order Now
Specification Test Results
Appearance Light yellow to light beige Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %