(+)-cis-Diltiazem hydrochloride 98%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: (2S,3S)-(+)-3-Acetoxy-2,3-dihydro-5-[2-(dimethylamino)ethyl]-2-(4-methoxyphenyl)-1,5-benzothiazepin-4(5H)-one Hydrochloride, Diltiazem Hydrochloride, (+)-cis-Diltiazem HCl, (+)-cis-3-Acetoxy-5-(2-dimethylaminoethyl)-2,3-dihydro-2-(4-methoxyphenyl)-1,5-benzothiazepin-4(5H)-one Hydrochloride, (2S,3S)-5-(2-(Dimethylamino)ethyl)-2-(4-methoxyphenyl)-4-oxo-2,3,4,5-tetrahydrobenzo[b][1,4]thiazepin-3-yl acetate hydrochloride
329080010 329080050 329080250
ACRO329080010EA 30.3 EUR
ACRO329080010 ACRO329080050 ACRO329080250
(+)-cis-Diltiazem hydrochloride 98%
(+)-cis-Diltiazem hydrochloride
Formula: C₂₂H₂₆N₂O₄S·HCl
MW: 450.98 g/mol
Melting Pt: 210…215 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00069252
CAS Number: 33286-22-5
Order Now
Specification Test Results
Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Specific optical rotation +110° to +120° (20°C, 589 nm) (c=1, H2O)