Ethyl-4-chloro-8-cyanoquinoline-3-carboxylate 97%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Ethyl 4-chloro-8-cyanoquinoline-3-carboxylate
449680010 449680050
ACRO449680010EA 120 EUR
ACRO449680010 ACRO449680050
Ethyl-4-chloro-8-cyanoquinoline-3-carboxylate 97%
Ethyl 4-chloro-8-cyanoquinoline-3-carboxylate
Formula: C₁₃H₉ClN₂O₂
MW: 260.68 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00173414
CAS Number: 77173-67-2
Order Now
Specification Test Results
Appearance Light yellow to light brown Powder
Infrared spectrum Conforms
Water ≤1 %
HPLC ≥96.0 %