Sulbactam
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Sulbactam acid, (2S,5R)-3,3-Dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid 4,4-Dioxide
455350050 455350250
ACRO455350050EA 73.3 EUR
ACRO455350050 ACRO455350250
Sulbactam
Sulbactam
Formula: C₈H₁₁NO₅S
MW: 233.24 g/mol
Melting Pt: 156 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00867005
CAS Number: 68373-14-8
Order Now
Specification Test Results
Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 146 to 151 °C
Water ≤0.5 %
HPLC ≥97.5 %