2-Heptyne 97%

Supplier: Thermo Scientific
Danger

455560250 455560050
ACRO455560250EA 139 EUR
ACRO455560250 ACRO455560050
2-Heptyne 97%
2-Heptyne
Formula: C₇H₁₂
MW: 96.17 g/mol
Boiling Pt: 110…111 °C (743 mmHg)
Density: 0.759 g/cm³
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00039970
CAS Number: 1119-65-9
UN: 3295
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4200 to 1.4230 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR