tert-Butyl methyl ether ACS
Supplier: Thermo Fisher Scientific
Danger
Synonyms: MTBE, 2-Methoxy-2-methylpropane, Methyl-t-Butyl Ether
447470010 447470025
ACRO447470010EA 47.5 EUR
ACRO447470010 ACRO447470025
tert-Butyl methyl ether ACS
tert-Butyl methyl ether
Formula: (CH₃)₃COCH₃
MW: 88.15 g/mol
Boiling Pt: 54…56 °C (760 mmHg)
Melting Pt: –110 °C
Density: 0.7404 g/cm³
Flash Pt: –28 °C
Storage Temperature: Ambient
MDL Number: MFCD00008812
CAS Number: 1634-04-4
UN: 2398
ADR: 3 II
Merck Index: 13,06059
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1712bc714ba0a06e0de37f7cf4e5117256f84425&vid=e1b314f829136075329c50f9e5a593c19ae28b4f&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Clear liquid
Water ≤0.05 %
GC ≥99.0 %
Color scale ≤10 APHA
Residue after evaporation ≤0.001 %
Peroxides ≤1 ppm