6-Methyl-3,5-heptadien-2-one 96%

Supplier: Thermo Scientific
Warning

Synonyms: 6-Methylhepta-3,5-dien-2-one

453210050 453210250 453211000
ACRO453210050EA 25.1 EUR
ACRO453210050 ACRO453210250 ACRO453211000
6-Methyl-3,5-heptadien-2-one 96%
6-Methyl-3,5-heptadien-2-one
Formula: C₈H₁₂O
MW: 124.18 g/mol
Boiling Pt: 168 °C
Density: 0.893 g/cm³
Flash Pt: 80 °C
Storage Temperature: Ambient
MDL Number: MFCD00043647
CAS Number: 1604-28-0

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5340 to 1.5360 (20°C, 589 nm)
GC ≥95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR