Ketoconazole 98%
Supplier: Thermo Scientific
Danger
Synonyms: (+/-)-cis-1-Acetyl-4-[4-[[2-(2,4-dichlorophenyl)-2-(imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazine, cis-1-Acetyl-4-[4-[[2-(2,4-dichlorophenyl)-2-(imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazine
455470010 455470050 455470250
ACRO455470010EA 62.8 EUR
ACRO455470010 ACRO455470050 ACRO455470250
Ketoconazole 98%
Ketoconazole
Formula: C₂₆H₂₈Cl₂N₄O₄
MW: 531.43 g/mol
Melting Pt: 148…150 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00058579
CAS Number: 65277-42-1
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Appearance White to light brown Powder
Infrared spectrum Conforms
Melting point 148 to 152 °C
Loss on drying ≤0.5 % (80 °C, 4 h) (vacuum)
HPLC ≥97.5 %