(1-Methyl-1H-pyrazol-5-yl)methylamine 97%
Supplier: Thermo Scientific
Danger
Synonyms: 5-(Aminomethyl)-1-methyl-1H-pyrazole
456030010 456030050
ACRO456030010EA 80.1 EUR
ACRO456030010 ACRO456030050
(1-Methyl-1H-pyrazol-5-yl)methylamine 97%
(1-Methyl-1H-pyrazol-5-yl)methylamine
Formula: C₅H₉N₃
MW: 111.15 g/mol
Boiling Pt: 140 °C (8 mmHg)
Storage Temperature: Refrigerator
MDL Number: MFCD03419808
CAS Number: 863548-52-1
UN: 2735
ADR: 8,III
Order Now
Specification Test Results
Appearance Clear colorless to yellow to brown Liquid
Infrared spectrum Conforms
Water ≤1.5 %
GC ≥96.0 %