Oleanolic acid 97%
Supplier: Thermo Fisher Scientific
Warning
456120050 456120250
ACRO456120050EA 72.9 EUR
ACRO456120050 ACRO456120250
Oleanolic acid 97%
Oleanolic acid
Formula: C₃₀H₄₈O₃
MW: 456.71 g/mol
Melting Pt: >300 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00064914
CAS Number: 508-02-1
Order Now
Specification Test Results
Appearance White to light yellow Powder
Infrared spectrum Conforms
Water ≤5 %
HPLC ≥96.0 %