2,4-Dimethyloxazole-5-carboxylic acid 97%
Supplier: Thermo Scientific
Warning
457350010 457350050
ACRO457350010EA 191 EUR
ACRO457350010 ACRO457350050
2,4-Dimethyloxazole-5-carboxylic acid 97%
2,4-Dimethyloxazole-5-carboxylic acid
Formula: C₆H₇NO₃
MW: 141.13 g/mol
Melting Pt: 229…231 °C
Storage Temperature: Ambient
MDL Number: MFCD06011090
CAS Number: 2510-37-4
Order Now
Specification Test Results
Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 225 to 235 °C
HPLC ≥96 %