Isothiazole 97%
Supplier: Thermo Fisher Scientific
Danger
458420010 458420050
ACRO458420010EA 163 EUR
ACRO458420010 ACRO458420050
Isothiazole 97%
Isothiazole
Formula: C₃H₃NS
MW: 85.12 g/mol
Flash Pt: 22 °C
Storage Temperature: Ambient
MDL Number: MFCD00020818
CAS Number: 288-16-4
UN: 1993
ADR: 3 II
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %