Econazole 99%
Supplier: Thermo Scientific
Warning
Synonyms: 1-(2-((4-Chlorobenzyl)oxy)-2-(2,4-dichlorophenyl)ethyl)-1H-imidazole, 1-[2,4-Dichloro-beta-[(p-chlorobenzyl)oxy]phenethyl]imidazole
458630050 458630250
ACRO458630050EA 51.5 EUR
ACRO458630050 ACRO458630250
Econazole 99%
Econazole
Formula: C₁₈H₁₅Cl₃N₂O
MW: 381.69 g/mol
Melting Pt: 91…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00800993
CAS Number: 27220-47-9
Order Now
Specification Test Results
Appearance White to off-white powder Powder
Infrared spectrum Conforms
Melting point 88 to 92 °C
Loss on drying ≤0.5 % (1 g, 60 °C, 4 h)
HPLC ≥98.5 %