3-(Piperidin-2-yl)pyridine 97%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: (±) Anabasine, 2-Pyridin-3-ylpiperidine
458980010 458980050
ACRO458980010EA 240 EUR
ACRO458980010 ACRO458980050
3-(Piperidin-2-yl)pyridine 97%
3-(Piperidin-2-yl)pyridine
Formula: C₁₀H₁₄N₂
MW: 162.236 g/mol
Boiling Pt: 270…272 °C (760 mmHg)
Melting Pt: 9 °C
Density: 1.046 g/cm³
Flash Pt: 93 °C
Storage Temperature: Freezer
MDL Number: MFCD00006370
CAS Number: 13078-04-1
UN: 3140
ADR: 6.1 II
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %