Methyl nervonate (15 cis), Nervonic acid methyl ester 99% analytical standard for GC
Supplier: Thermo Fisher Scientific
Synonyms: Methyl 15(Z)-tetracosenoate, 15(Z)-Tetracosenoic acid methyl ester, Methyl nervonate (15 cis), Nervonic acid methyl ester
460371000 460375000
ACRO460371000EA 72.1 EUR
ACRO460371000 ACRO460375000
Methyl nervonate (15 cis), Nervonic acid methyl ester 99% analytical standard for GC
Methyl nervonate
Formula: C₂₅H₄₈O₂
Storage Temperature: Freezer
MDL Number: MFCD00056324
CAS Number: 2733-88-2
Order Now
Specification Test Results
Appearance White low melting solid
Infrared spectrum Conforms
GC ≥98.5 %