Procarbazine hydrochloride
Supplier: Thermo Scientific
Danger
Synonyms: N-Isopropyl-4-[(2-methylhydrazino)methyl]benzamide, N-Isopropyl-4-((2-methylhydrazinyl)methyl)benzamide hydrochloride, Procarbazine HCl
461180010 461182500
ACRO461180010EA 466 EUR
ACRO461180010 ACRO461182500
Procarbazine hydrochloride
Procarbazine hydrochloride
Formula: C₁₂H₁₉N₃O·HCl
MW: 257.76 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00072082
CAS Number: 366-70-1
Order Now
Specification Test Results
Appearance White to tan Powder
Infrared spectrum Conforms
Loss on drying ≤2 %
HPLC ≥97.5 %