1H-Imidazole-4-carbonitrile 95%
Supplier: Thermo Scientific
Danger
461360010 461360050
ACRO461360010EA 60.7 EUR
ACRO461360010 ACRO461360050
1H-Imidazole-4-carbonitrile 95%
1H-Imidazole-4-carbonitrile
Formula: C₄H₃N₃
MW: 93.09 g/mol
Melting Pt: 32…35 °C
Storage Temperature: Ambient
MDL Number: MFCD00233358
CAS Number: 57090-88-7
UN: 3439
ADR: 6.1,III
Order Now
Specification Test Results
Appearance White to yellow to brown Powder
Infrared spectrum Conforms
Melting point 141 to 148 °C
Water ≤2.5 %
HPLC ≥94 %