Anti-CASP3 Polyclonal Antibody

Supplier: Boster
BSBTPB9188EA 550 EUR
BSBTPB9188
Anti-CASP3 Polyclonal Antibody
Antibodies
Polyclonal antibody for CASPASE 3/CASP3 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. CASPASE 3/CASP3 information: Molecular Weight: 31608 MW; Subcellular Localization: Cytoplasm; Tissue Specificity: Highly expressed in lung, spleen, heart, liver and kidney. Moderate levels in brain and skeletal muscle, and low in testis. Also found in many cell lines, highest expression in cells of the immune system.

Type: Primary
Antigen: CASP3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR