VWR® Anti-Evaporation Spheres for Water Baths
Supplier: VWR Collection
462-7050EA 131 EUR
462-7050 462-7049 462-0584
VWR® Anti-Evaporation Spheres for Water Baths
Beads Plastic Beads
Lightweight, economic PP spheres which form a blanket on the liquid surface. Spheres have 20 mm Ø, 275 spheres cover approximately 1000 cm².
  • Reduce heat loss by 77%, and evaporation by 87%
  • Insulate like a solid, and still allow instant access to the liquid in the bath