Jars, without cap
Supplier: RIXIUS
4-1101-0025-25 4-1101-0350-90 4-1201-0050-35 4-1201-0100-45 4-1201-0325-65 4-1201-0500-90
RIXI4-1101-0025-25EA 826 EUR
RIXI4-1101-0025-25 RIXI4-1101-0350-90 RIXI4-1201-0050-35 RIXI4-1201-0100-45 RIXI4-1201-0325-65 RIXI4-1201-0500-90
Jars, without cap
Jars General Use Jars
These slip lid jars are made of white PP.
  • Very good chemical resistance
  • Very good temperature resistance
Informace pro objednávání: Caps need to be ordered seperately.
Upozornení: *The product(s) on this page are outside the scope of the EU Directive 2019/904