Sodium chloride-³⁵Cl 99 atom % ³⁵Cl, Sigma-Aldrich®

Supplier: Merck
Warning

451940-100MG 451940-250MG
SIAL451940-100MGEA 3030 EUR
SIAL451940-100MG SIAL451940-250MG
Sodium chloride-³⁵Cl 99 atom % ³⁵Cl, Sigma-Aldrich®
Sodium chloride-³⁵Cl
Formula: Na³⁵Cl
Melting Pt: 801 °C (lit.)
MDL Number: MFCD00198119
CAS Number: 20510-55-8

Order Now

Specification Test Results

Isotopic purity 99 atom% ³⁵Cl
Mass shift Depleted


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR