Restriction enzymes, EcoRI, FastDigest™, Fermentas
Supplier: Thermo Fisher Scientific
FD0274 FD0275
FD0274EA 63.75 EUR
FD0274 FD0275
Restriction enzymes, EcoRI, FastDigest™, Fermentas
Nucleic Acid Reagents Restriction Enzymes
Thermo Scientific™ FastDigest™ enzymes are an advanced line of restriction enzymes for rapid DNA digestion.
  • Activity (100%) of all FastDigest™ enzymes in one universal reaction buffer
  • Direct loading on gels with FastDigest™ Green buffer
  • Buffer compatibility (100%) with downstream applications
Designed to complete digestion in 5 to 15 minutes.
Order Now
SPECIFICATIONS
Environmentally Preferable
Application
Enzyme name EcoRI
Source
Overhang type 5' overhang
Compatible buffer FastDigest Green Buffer or FastDigest Buffer (colorless)