Bottles, narrow neck, PYREX®
Supplier: SciLabware
1526/16D 1526/22D 1526/18D 1526/20D
201-0010EA 146 EUR
201-0010 BIBB1526/22D BIBB1526/18D BIBB1526/20D
Bottles, narrow neck, PYREX®
Bottles Glass Bottles
PYREX®, borosilicate glass.
  • For use with rubber stoppers
  • Excellent chemical resistance, high temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Marked with volume and equipped with a white labelling field.
Certifikace: ISO 4796.