Anti-12(L)-HETE Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
ENZOADI905053100EA 388.3 EUR
ENZOADI905053100 ALEXADI905053100
Anti-12(L)-HETE Rabbit Polyclonal Antibody
Antibodies
HETEs are byproducts generated by the metabolism of arachidonic acid by lipoxygenases. 12(S)-HETE is the stereospecific hydroxy product from the reduction of 12(S)-hydroperoxy tetraenoic eicosatetraenoic acid [12(S)-HpETE], which itself is a 12-lipoxygenase metabolite of arachidonic acid. 12(S)-HETE is chemotactic and chemokinetic for polymorphonuclear leukocytes and vascular smooth muscle cells. It also acts as a second messenger in angiotensin-II induced aldosterone production. 15-hydroxyeicosatetraenoic acid [15(S)-HETE] is the major hydroxy derivative of arachidonic acid when acted upon by 15-lipoxygenase. It is also the primary monohydroxy acid synthesized by the lipoxygenase activity of Cyclooxygenase-1 upon linoleic acid. 15(S)-HETE acts as a paracrine regulator of smooth muscle and lung neutrophil recruitment, and elevated levels of 15(S)-HETE are associated with asthma, rhinitis, chronic paranasal sinusitis and rheumatoid arthritis.

  • For ELISA, Flow Cytometry

Species independent.

Type: Primary
Antigen: 12(L)-HETE
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: pan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR