Anti-IL33 Mouse Monoclonal Antibody
Supplier: Enzo Life Sciences
ENZOALX804840C100EA 517 EUR
ENZOALX804840C100 ALEXALX804840C100
Anti-IL33 Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG1
  • For ELISA, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot
Recognizes human, mouse and rat pro-IL-33 (precursor) and mature IL-33.
Type: Primary
Antigen: IL33
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human