Anti-JNK1 Rabbit Polyclonal Antibody
Supplier: ENZO LIFE SCIENCES
BMLSA2720020 BMLSA2720100
ENZOBMLSA2720020EA 198 EUR
ENZOBMLSA2720020 ENZOBMLSA2720100
Anti-JNK1 Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit
  • For Immunohistochemistry, Western Blot
Recognizes human and rat JNK 1&2 dually phosphorylated at Thr183 and Tyr185. Does not cross-react with unphosphorylated JNK1&2.
Type: Primary
Antigen: JNK1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat
New Product
Order Now
SPECIFICATIONS
Antigen symbol JNK1 JNK1
Antigen name c-Jun N-terminal kinase 1 c-Jun N-terminal kinase 1
Environmentally Preferable
Antigen synonyms JNK1,SARP|JNK1A2|Stress activated protein kinase|JNK 1|JNK|JAK 1A|Mitogen-activated protein kinase 8|JNK1|MAPK8|JAK1A JNK1,SARP|JNK1A2|Stress activated protein kinase|JNK 1|JNK|JAK 1A|Mitogen-activated protein kinase 8|JNK1|MAPK8|JAK1A
Antibody type Primary Primary
Clonality Polyclonal Polyclonal
Reactivity Human,Rat
Host Rabbit Rabbit
Isotype
Western blot Yes Yes
ImmunoChemistry Yes Yes
Form Liquid, in PBS containing BSA and sodium azide Liquid, in PBS containing BSA and sodium azide