Anti-PRAS40 Rabbit Polyclonal Antibody
Supplier: Enzo Life Sciences
ALEXBML-SA360-0020EA 233 EUR
ALEXBML-SA360-0020 ALEXBML-SA360-0100 ENZOBMLSA3600100
Anti-PRAS40 Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit
  • For Western Blot
Recognizes PRAS40 phosphorylated at pThr246.
Type: Primary
Antigen: PRAS40
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: pan