Anti-GFP Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
ENZOALX210199R100EA 551.1 EUR
ENZOALX210199R100 ALEXALX210199R100
Anti-GFP Rabbit Polyclonal Antibody
Antibodies
Green Fluorescent Protein (GFP) is responsible for bioluminescence in the jellyfish Aequorea victoria. This protein has quickly become a powerful research tool for assessing gene expression and subcellular protein distribution in fixed or living cells. GFP is excited by and brightly fluoresces when exposed to UV or blue light. This feature makes it ideal as a marker for use in fluorescence microscopy, cytometry, tagging fusion proteins, and assaying transcriptional regulation from gene promoters in vivo. Numerous GFP variants with enhanced and shifted emission spectra (blue, green, and yellow) have been developed through aa substitutions at specific residues.

Type: Primary
Antigen: GFP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Species independent
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR