Anti-Biotin Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
ALEXENZ-32892-6000EA 685 EUR
ALEXENZ-32892-6000 ENZOENZ328926000
Anti-Biotin Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Biotin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Species independent
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR