Anti-APP Mouse Monoclonal Antibody [clone: 11A50-B10]
Supplier: ENZO LIFE SCIENCES
ENZABS1160200 ENZABS1161000
ENZOENZABS1160200EA 838 EUR
ENZOENZABS1160200 ENZOENZABS1161000
Anti-APP Mouse Monoclonal Antibody [clone: 11A50-B10]
Antibodies
Recommended Applications: ELISA, IHC, WB
Species reactivity: Human, Mouse
Type: Primary
Antigen: APP
Clonality: Monoclonal
Clone: 11A50-B10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse