Testosterone ELISA kit
Supplier: Enzo Life Sciences
ALEXADI-900-065EA 570 EUR
ALEXADI-900-065 ENZOADI900065
Testosterone ELISA kit
Assays ELISAs
For the quantitative determination of testosterone in culture supernatants, plasma, serum, and saliva from any species. Not suitable for human serum and plasma. For those samples use product 89133-298. Cited sample types include tissue and feces.
The Testosterone EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 3 hours. Absorbance is read at 405 nm. Ready-to-use liquid color-coded reagents are provided to reduce error. The reproducible results lot-after-lot provide excellent consistency, ideal for longitudinal studies.
Informace pro objednávání: Contains GxM IgG Microtiter plate, Conjugate, Antibody, Assay buffer 3, Wash buffer concentrate, Standard, Steroid displacement reagent, pNpp Substrate, Stop solution. Alternative size available: 89141-308 (5x96 wells)