RANKL (total), soluble (human) ELISA kit
Supplier: Enzo Life Sciences
ALEXALX850019-KI01EA 1640 EUR
ALEXALX850019-KI01 ENZOALX850019KI01
RANKL (total), soluble (human) ELISA kit
Assays ELISAs
For the quantitative determination of free and OPG-bound human soluble RANKL from serum, plasma and cell culture supernatant.