FSH ELISA kit, Grow Cells
Supplier: Enzo Life Sciences
ENZOENZKIT1080001EA 295.9 EUR
ENZOENZKIT1080001 ALEXENZKIT1080001
FSH ELISA kit, Grow Cells
Assays ELISAs
Highly sensitive FSH ELISA kit enabling detection of FSH in serum, plasma and tissue culture media in just 2 hours.
  • Sensitive measurement of FSH, detecting as little as 0,5 mIU/ml
  • Negligible reactivity from similar glycoprotein hormones
  • High throughput format with results in two hours for up to 37 samples in duplicate
  • Fully quantitative results that surpass semi-quantitative Western blot analysis
For the quantitative determination of FSH in tissue culture media, serum and plasma in human samples.
The FSH ELISA kit is a colorimetric sandwich immunoassay kit with results in two hours.
Informace pro objednávání: Contains Microtiter Plate, Assay Buffer 13, Standard, Wash Buffer Concentrate, Antibody, Tracer, Conjugate, Substrate, and Stop Solution.