Anti-IgG Sheep Polyclonal Antibody (Biotin)

Supplier: Immunoreagents
IMMRSH-003-CBIOEA 98.5 EUR
IMMRSH-003-CBIO
Anti-IgG Sheep Polyclonal Antibody (Biotin)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Sheep
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR